siRNA Details
Gene Symbol: CELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033342.1 | 1, 2 | 135 | |||
XM_006722837.3 | XP_006722900.1 | 1, 2 | 236 | |||
GenBank mRNA | AK294109.1 | - | 1, 2 | 135 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033342.1 | 1, 2 | 135 | |||
XM_006722837.3 | XP_006722900.1 | 1, 2 | 236 | |||
GenBank mRNA | AK294109.1 | - | 1, 2 | 135 |