siRNA Details
| Gene Symbol: LINC00671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027254.1 | 1 | 91 | |||
| GenBank mRNA | AK055784.1 | - | 1 | 61 | ||
| AK298293.1 | - | 1 | 91 | |||
| BC122867.1 | - | 1 | 13 | |||
| BC122868.1 | - | 1 | 13 | |||
| DC361857.1 | - | 1 | 91 | |||

LINC00671
Genomic Map