siRNA Details
Gene Symbol: LINC00671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027254.1 | 2 | 157 | |||
GenBank mRNA | AK055784.1 | - | 3 | 359 | ||
BC122867.1 | - | 2 | 79 | |||
BC122868.1 | - | 2 | 79 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027254.1 | 2 | 157 | |||
GenBank mRNA | AK055784.1 | - | 3 | 359 | ||
BC122867.1 | - | 2 | 79 | |||
BC122868.1 | - | 2 | 79 |