siRNA Details
| Gene Symbol: AATK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027255.1 | 2 | 379 | |||
| GenBank mRNA | AK126811.1 | - | 2 | 379 | ||
| BC140737.1 | - | 2 | 370 | |||
| BC171777.1 | - | 2 | 370 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA135843.1 | - | 2 | 379 | Chr. 17 - 81165507 - 81179051 [+] on Build GRCh38 | |

AATK-AS1
Genomic Map