siRNA Details
| Gene Symbol: AATK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027255.1 | 3 | 561 | |||
| GenBank mRNA | AK126811.1 | - | 3 | 561 | ||
| BC140737.1 | - | 3 | 552 | |||
| BC171777.1 | - | 3 | 552 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AATK-AS1| Gene Symbol: AATK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027255.1 | 3 | 561 | |||
| GenBank mRNA | AK126811.1 | - | 3 | 561 | ||
| BC140737.1 | - | 3 | 552 | |||
| BC171777.1 | - | 3 | 552 | |||