siRNA Details
| Gene Symbol: LINC01366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026945.2 | 3 | 1834 | |||
| NR_108024.1 | 3 | 1815 | ||||
| NR_108025.1 | 2 | 1744 | ||||
| GenBank mRNA | AK093725.1 | - | 3 | 2530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01366| Gene Symbol: LINC01366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026945.2 | 3 | 1834 | |||
| NR_108024.1 | 3 | 1815 | ||||
| NR_108025.1 | 2 | 1744 | ||||
| GenBank mRNA | AK093725.1 | - | 3 | 2530 | ||