siRNA Details
| Gene Symbol: LOC100240735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026658.1 | 2 | 215 | |||
| GenBank mRNA | CN361140.1 | - | 2 | 215 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100240735| Gene Symbol: LOC100240735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026658.1 | 2 | 215 | |||
| GenBank mRNA | CN361140.1 | - | 2 | 215 | ||