siRNA Details
| Gene Symbol: LINC01119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024452.1 | 3 | 334 | |||
| GenBank mRNA | AK092497.1 | - | 3 | 1281 | ||
| BC017573.1 | - | 3 | 345 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01119| Gene Symbol: LINC01119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024452.1 | 3 | 334 | |||
| GenBank mRNA | AK092497.1 | - | 3 | 1281 | ||
| BC017573.1 | - | 3 | 345 | |||