siRNA Details
| Gene Symbol: EGFEM1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021485.2 | 16 | 2782 | |||
| GenBank mRNA | BC011266.2 | - | 6 | 1078 | ||
| BC103698.1 | - | 4 | 899 | |||
| BC104748.1 | - | 4 | 898 | |||
| BC105690.1 | - | 4 | 898 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EGFEM1P| Gene Symbol: EGFEM1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021485.2 | 16 | 2782 | |||
| GenBank mRNA | BC011266.2 | - | 6 | 1078 | ||
| BC103698.1 | - | 4 | 899 | |||
| BC104748.1 | - | 4 | 898 | |||
| BC105690.1 | - | 4 | 898 | |||