siRNA Details
| Gene Symbol: RBMY2EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001574.2 | 4 | 325 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U94388.3 | - | 4 | 325 | Chr. Y - 21395148 - 21401562 [-] on Build GRCh38 | |

RBMY2EP
Genomic Map