siRNA Details
| Gene Symbol: RPSAP58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003662.3 | 2 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPSAP58| Gene Symbol: RPSAP58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003662.3 | 2 | 205 | |||