siRNA Details
| Gene Symbol: LOC93622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015433.1 | 2 | 768 | |||
| GenBank mRNA | AK026375.1 | - | 1 | 1072 | ||
| BC006130.2 | - | 2 | 768 | |||
| BC021542.1 | - | 1 | 1044 | |||
| BC026970.1 | - | 2 | 774 | |||
| BC040997.1 | - | 2 | 1415 | |||
| BC066911.1 | - | 1 | 1152 | |||

LOC93622
Genomic Map