siRNA Details
Gene Symbol: RBMY2FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002193.2 | 4 | 2007 | |||
GenBank mRNA | BC026077.2 | - | 4 | 2009 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RBMY2FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002193.2 | 4 | 2007 | |||
GenBank mRNA | BC026077.2 | - | 4 | 2009 |