siRNA Details
| Gene Symbol: CEACAM22P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027754.2 | 1 | 78 | |||
| GenBank mRNA | AK123850.1 | - | 1 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEACAM22P| Gene Symbol: CEACAM22P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027754.2 | 1 | 78 | |||
| GenBank mRNA | AK123850.1 | - | 1 | 78 | ||