siRNA Details
| Gene Symbol: ST7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002330.1 | 1 | 1454 | |||
| GenBank mRNA | AF400039.1 | - | LIT2006 ST7OT1 |
1 | 1454 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ST7-AS1| Gene Symbol: ST7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002330.1 | 1 | 1454 | |||
| GenBank mRNA | AF400039.1 | - | LIT2006 ST7OT1 |
1 | 1454 | |