siRNA Details
| Gene Symbol: ST7-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002331.3 | 6 | 611 | |||
| NR_109981.1 | 5 | 507 | ||||
| GenBank mRNA | AF400045.1 | - | LIT1899 ST7OT2 |
6 | 601 | |
| DB054726.1 | - | 5 | 507 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX097853.1 | - | 6 | 578 | Chr. 7 - 117112518 - 117145559 [-] on Build GRCh38 | |

ST7-AS2
Genomic Map