siRNA Details
| Gene Symbol: ST7-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002331.3 | 6 | 894 | |||
| NR_109981.1 | 5 | 790 | ||||
| GenBank mRNA | AF400045.1 | - | LIT1899 ST7OT2 |
6 | 883 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ST7-AS2| Gene Symbol: ST7-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002331.3 | 6 | 894 | |||
| NR_109981.1 | 5 | 790 | ||||
| GenBank mRNA | AF400045.1 | - | LIT1899 ST7OT2 |
6 | 883 | |