siRNA Details
| Gene Symbol: RPL23AP87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029406.1 | 3 | 1339 | |||
| GenBank mRNA | AK125669.1 | - | 3 | 1337 | ||
| Gene Symbol: LOC728323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125248.1 | - | 7 | 1632 | ||

RPL23AP87
Genomic Map