siRNA Details
| Gene Symbol: LINC01106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027244.1 | 1 | 178 | |||
| GenBank mRNA | BP341570.1 | - | 1 | 164 | ||
| CR749351.1 | - | 1 | 84 | |||
| DB205082.1 | - | 1 | 178 | |||
| Gene Symbol: LINC01123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046110.1 | 1 | 167 | |||
| NR_046111.1 | 1 | 167 | ||||
| NR_046112.1 | 1 | 167 | ||||
| GenBank mRNA | AK056084.1 | - | 1 | 84 | ||
| BP247062.1 | - | 1 | 167 | |||
| DA999847.1 | - | 1 | 98 | |||

LINC01106
Genomic Map