siRNA Details
| Gene Symbol: HPYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026684.1 | 1 | 236 | |||
| GenBank mRNA | AF200341.1 | - | 1 | 236 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPYR1| Gene Symbol: HPYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026684.1 | 1 | 236 | |||
| GenBank mRNA | AF200341.1 | - | 1 | 236 | ||