siRNA Details
| Gene Symbol: TMEM51-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027136.1 | 1 | 42 | |||
| GenBank mRNA | AK074283.1 | - | 1 | 44 | ||
| BC128126.1 | - | 1 | 24 | |||
| BC128127.1 | - | 1 | 24 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM51-AS1| Gene Symbol: TMEM51-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027136.1 | 1 | 42 | |||
| GenBank mRNA | AK074283.1 | - | 1 | 44 | ||
| BC128126.1 | - | 1 | 24 | |||
| BC128127.1 | - | 1 | 24 | |||