siRNA Details
| Gene Symbol: MTMR9LP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026850.1 | 7 | 2168 | |||
| GenBank mRNA | AK055560.1 | - | 8 | 1900 | ||
| AK096109.1 | - | 6 | 1720 | |||
| AL833686.1 | - | 9 | 2468 | |||
| BC016171.1 | - | 4 | 1013 | |||
| BC041632.1 | - | 4 | 823 | |||

MTMR9LP
Genomic Map