siRNA Details
| Gene Symbol: MTMR9LP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026850.1 | 2 | 984 | |||
| GenBank mRNA | AK055560.1 | - | 3 | 798 | ||
| AK096109.1 | - | 1 | 536 | |||
| AL833686.1 | - | 2 | 894 | |||
| BC016171.1 | - | 1 | 168 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTMR9LP| Gene Symbol: MTMR9LP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026850.1 | 2 | 984 | |||
| GenBank mRNA | AK055560.1 | - | 3 | 798 | ||
| AK096109.1 | - | 1 | 536 | |||
| AL833686.1 | - | 2 | 894 | |||
| BC016171.1 | - | 1 | 168 | |||