siRNA Details
| Gene Symbol: ZNF678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033184.1 | 3 | 370 | |||
| NR_102302.1 | 4 | 463 | ||||
| GenBank mRNA | AK307248.1 | - | 4 | 439 | ||
| BC042500.1 | - | 3 | 346 | 4823179 | ||
| DA213851.1 | - | 3 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF678| Gene Symbol: ZNF678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033184.1 | 3 | 370 | |||
| NR_102302.1 | 4 | 463 | ||||
| GenBank mRNA | AK307248.1 | - | 4 | 439 | ||
| BC042500.1 | - | 3 | 346 | 4823179 | ||
| DA213851.1 | - | 3 | 371 | |||