siRNA Details
Gene Symbol: ZNF678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033184.1 | 3, 4 | 457 | |||
NR_102302.1 | 4, 5 | 550 | ||||
GenBank mRNA | AK307248.1 | - | 4, 5 | 526 | ||
BC042500.1 | - | 3, 4 | 433 | 4823179 | ||
DA213851.1 | - | 3, 4 | 458 |
