siRNA Details
| Gene Symbol: ZNF678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033184.1 | 2, 3 | 341 | |||
| NR_102302.1 | 3, 4 | 434 | ||||
| GenBank mRNA | AK307248.1 | - | 3, 4 | 410 | ||
| BC042500.1 | - | 2, 3 | 317 | 4823179 | ||
| DA213851.1 | - | 2, 3 | 342 | |||

ZNF678
Genomic Map