siRNA Details
Gene Symbol: ZNF678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033184.1 | 1, 2 | 207 | |||
GenBank mRNA | BC042500.1 | - | 1, 2 | 183 | 4823179 | |
DA213851.1 | - | 1, 2 | 208 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033184.1 | 1, 2 | 207 | |||
GenBank mRNA | BC042500.1 | - | 1, 2 | 183 | 4823179 | |
DA213851.1 | - | 1, 2 | 208 |