siRNA Details
| Gene Symbol: PLK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243079.1 | NP_001230008.1 | 12 | 1359 | ||
| GenBank mRNA | AK054808.1 | - | 12 | 1359 | ||
| AK122648.1 | - | 11 | 1531 | |||
| BC100791.2 | - | 9 | 923 | |||
| BC100792.1 | - | 9 | 923 | |||
| DQ424898.1 | - | 6 | 643 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI827807.1 | - | 6 | 574 | Chr. 19 - 1528003 - 1535429 [+] on Build GRCh38 | |

PLK5
Genomic Map