siRNA Details
| Gene Symbol: PLK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243079.1 | NP_001230008.1 | 4 | 442 | ||
| GenBank mRNA | AI651075.1 | - | 4 | 400 | ||
| AK054808.1 | - | 4 | 442 | |||
| AK122648.1 | - | 3 | 614 | |||
| BF364682.1 | - | 2 | 121 | |||
| DA058511.1 | - | 4 | 439 | |||

PLK5
Genomic Map