siRNA Details
| Gene Symbol: PLK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243079.1 | NP_001230008.1 | 6 | 663 | ||
| GenBank mRNA | AK054808.1 | - | 6 | 663 | ||
| AK122648.1 | - | 5 | 835 | |||
| BC100791.2 | - | 3 | 227 | |||
| BC100792.1 | - | 3 | 227 | |||
| BC100793.2 | - | 3 | 227 | |||

PLK5
Genomic Map