siRNA Details
| Gene Symbol: CFL1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028492.1 | 4 | 1586 | |||
| GenBank mRNA | BC031631.1 | - | 4 | 1586 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFL1P1| Gene Symbol: CFL1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028492.1 | 4 | 1586 | |||
| GenBank mRNA | BC031631.1 | - | 4 | 1586 | ||