siRNA Details
| Gene Symbol: LINC00661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026828.1 | 9 | 2585 | |||
| GenBank mRNA | AK128240.1 | - | 4 | 2105 | ||
| CR749400.1 | - | 9 | 2596 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00661| Gene Symbol: LINC00661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026828.1 | 9 | 2585 | |||
| GenBank mRNA | AK128240.1 | - | 4 | 2105 | ||
| CR749400.1 | - | 9 | 2596 | |||