siRNA Details
| Gene Symbol: LINC00661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026828.1 | 9 | 2934 | |||
| GenBank mRNA | AK128240.1 | - | 4 | 2454 | ||
| CR749400.1 | - | 9 | 2945 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI827405.1 | - | 5 | 718 | Chr. 19 - 16021928 - 16027109 [+] on Build GRCh38 | |

LINC00661
Genomic Map