siRNA Details
| Gene Symbol: LOC339568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027124.1 | 4 | 584 | |||
| GenBank mRNA | BC043524.1 | - | 4 | 584 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830782.1 | - | 4 | 585 | Chr. 20 - 39214614 - 39224748 [-] on Build GRCh38 | |

LOC339568
Genomic Map