siRNA Details
| Gene Symbol: LINC00161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026552.1 | 2 | 681 | |||
| NR_026553.1 | 2 | 325 | ||||
| GenBank mRNA | AY063458.1 | - | 2 | 681 | ||
| AY063459.1 | - | 2 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00161| Gene Symbol: LINC00161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026552.1 | 2 | 681 | |||
| NR_026553.1 | 2 | 325 | ||||
| GenBank mRNA | AY063458.1 | - | 2 | 681 | ||
| AY063459.1 | - | 2 | 325 | |||