siRNA Details
Gene Symbol: LINC00161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026552.1 | 2 | 795 | |||
NR_026553.1 | 2 | 439 | ||||
GenBank mRNA | AY063458.1 | - | 2 | 795 | ||
AY063459.1 | - | 2 | 439 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026552.1 | 2 | 795 | |||
NR_026553.1 | 2 | 439 | ||||
GenBank mRNA | AY063458.1 | - | 2 | 795 | ||
AY063459.1 | - | 2 | 439 |