siRNA Details
| Gene Symbol: PCAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026555.1 | 2 | 474 | |||
| GenBank mRNA | AF388200.1 | - | 2 | 469 | ||
| AF462307.1 | - | 2 | 424 | |||
| AK056825.1 | - | 2 | 474 | |||
| BC105591.1 | - | 2 | 572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCAT4| Gene Symbol: PCAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026555.1 | 2 | 474 | |||
| GenBank mRNA | AF388200.1 | - | 2 | 469 | ||
| AF462307.1 | - | 2 | 424 | |||
| AK056825.1 | - | 2 | 474 | |||
| BC105591.1 | - | 2 | 572 | |||