siRNA Details
| Gene Symbol: LINC00493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015432.1 | 1, 2 | 168 | |||
| GenBank mRNA | AK024594.1 | - | 1, 2 | 168 | ||
| BC015983.1 | - | 1, 2 | 163 | |||
| BC150559.1 | - | 1, 2 | 150 | |||
| BC150560.1 | - | 1, 2 | 150 | |||

LINC00493
Genomic Map