siRNA Details
| Gene Symbol: LINC00493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015432.1 | 2 | 464 | |||
| GenBank mRNA | AF147354.1 | - | 2 | 442 | ||
| AK024594.1 | - | 2 | 464 | |||
| BC015983.1 | - | 2 | 459 | |||
| BC039422.1 | - | 2 | 443 | |||
| BC092494.1 | - | 2 | 465 | |||

LINC00493
Genomic Map