siRNA Details
| Gene Symbol: CCDC163P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033296.1 | 5 | 1977 | |||
| NR_033297.1 | 5 | 2020 | ||||
| NR_033298.1 | 6 | 2105 | ||||
| GenBank mRNA | AK054714.1 | - | 5 | 1989 | ||
| BC036618.2 | - | 5 | 1637 | |||
| BC047421.1 | - | 6 | 2074 | |||

CCDC163P
Genomic Map