siRNA Details
| Gene Symbol: CCDC163P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033296.1 | 5 | 1329 | |||
| NR_033297.1 | 5 | 1372 | ||||
| NR_033298.1 | 6 | 1457 | ||||
| GenBank mRNA | AK054714.1 | - | 5 | 1341 | ||
| BC036618.2 | - | 5 | 990 | |||
| BC047421.1 | - | 6 | 1426 | |||

CCDC163P
Genomic Map