siRNA Details
| Gene Symbol: LINC00634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024355.1 | 2 | 1464 | |||
| GenBank mRNA | BC040153.1 | - | 2 | 1353 | ||
| BC068568.1 | - | 2 | 1464 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00634| Gene Symbol: LINC00634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024355.1 | 2 | 1464 | |||
| GenBank mRNA | BC040153.1 | - | 2 | 1353 | ||
| BC068568.1 | - | 2 | 1464 | |||