siRNA Details
| Gene Symbol: ZNF497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CA438336.1 | - | 1 | 496 | ||
| Gene Symbol: A1BG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015380.2 | 4 | 1783 | |||
| GenBank mRNA | AK027222.1 | - | 4 | 1783 | ||
| BC006144.1 | - | 1 | 615 | |||
| BC040926.1 | - | 3 | 1371 | |||

ZNF497
Genomic Map