siRNA Details
| Gene Symbol: KPNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026698.1 | 6 | 727 | |||
| XM_005247439.3 | XP_005247496.1 | 3 | 1234 | |||
| XM_011512797.2 | XP_011511099.1 | 6 | 2201 | |||
| XM_017006363.1 | XP_016861852.1 | 6 | 2175 | |||
| XM_017006364.1 | XP_016861853.1 | 6 | 604 | |||
| GenBank mRNA | AK125626.1 | - | 2 | 554 | ||
| AK128280.1 | - | 3 | 1234 | |||
| AK299030.1 | - | 5 | 613 | |||
| AK312713.1 | - | 6 | 722 | |||
| DN991078.1 | - | 6 | 569 | |||

KPNA1
Genomic Map