siRNA Details
| Gene Symbol: LINC00608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015390.1 | 2 | 913 | |||
| NR_024385.1 | 2 | 1389 | ||||
| GenBank mRNA | AK098539.1 | - | 2 | 1389 | ||
| BC049373.1 | - | 2 | 913 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432733.1 | - | 2 | 913 | Chr. 2 - 218976284 - 218979660 [+] on Build GRCh38 | |

LINC00608
Genomic Map