siRNA Details
| Gene Symbol: BMS1P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC008503.1 | - | 1 | 132 | ||
| Gene Symbol: BMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005271846.3 | XP_005271903.1 | 1 | 136 | ||

BMS1P6
Genomic Map