siRNA Details
| Gene Symbol: MYADML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003143.2 | 1 | 1492 | |||
| GenBank mRNA | AK057470.1 | - | 1 | 1479 | ||
| BC031088.1 | - | 1 | 1482 | |||
| BC038980.1 | - | 1 | 1449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYADML| Gene Symbol: MYADML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003143.2 | 1 | 1492 | |||
| GenBank mRNA | AK057470.1 | - | 1 | 1479 | ||
| BC031088.1 | - | 1 | 1482 | |||
| BC038980.1 | - | 1 | 1449 | |||