siRNA Details
| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 4, 5 | 374 | |||
| NR_028410.1 | 3, 4 | 343 | ||||
| NR_028411.1 | 3, 4 | 307 | ||||
| GenBank mRNA | BC144506.1 | - | 3, 4 | 331 | ||
| BC144508.1 | - | 4, 5 | 362 | |||
| BU585030.1 | - | 3, 4 | 326 | |||

ENSG00000187012
Genomic Map