siRNA Details
| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 1 | 49 | |||
| NR_028410.1 | 1 | 49 | ||||
| NR_028411.1 | 1 | 49 | ||||
| GenBank mRNA | BC144506.1 | - | 1 | 37 | ||
| BC144508.1 | - | 1 | 37 | |||
| BU585030.1 | - | 1 | 68 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00207| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 1 | 49 | |||
| NR_028410.1 | 1 | 49 | ||||
| NR_028411.1 | 1 | 49 | ||||
| GenBank mRNA | BC144506.1 | - | 1 | 37 | ||
| BC144508.1 | - | 1 | 37 | |||
| BU585030.1 | - | 1 | 68 | |||