siRNA Details
| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 4 | 272 | |||
| NR_028410.1 | 3 | 241 | ||||
| NR_028411.1 | 3 | 205 | ||||
| GenBank mRNA | BC144506.1 | - | 3 | 229 | ||
| BC144508.1 | - | 4 | 260 | |||
| BU585030.1 | - | 3 | 224 | |||

LINC00207
Genomic Map